CAS RN 333753-67-6
CAS RN 333753-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-[(2R)-3-[[3-Fluoro-4-(4-morpholinyl)phenyl]amino]-2-hydroxypropyl]acetamide, TRC
CAS: 333753-67-6 Molecular Formula: C15H22FN3O3 Molecular Weight (g/mol): 311.35 Synonym: Acetamide, N-[(2R)-3-[[3-fluoro-4-(4-morpholinyl)phenyl]amino]-2-hydroxypropyl]-,N-[(2R)-3-(3-Fluoro-4-morpholinoanilino)-2-hydroxypropyl]acetamide IUPAC Name: N-[(2R)-3-(3-fluoro-4-morpholin-4-ylanilino)-2-hydroxypropyl]acetamide SMILES: CC(=O)NC[C@H](O)CNc1ccc(N2CCOCC2)c(F)c1
2
N-[(2R)-3-(3-Fluoro-4-morpholinoanilino)-2-hydroxypropyl]acetamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.