accessibility menu, dialog, popup

UserName

OHNOHNFFF

CAS RN 3335-46-4

IUPAC Name: 2-hydroxy-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carbonitrile
Synonyms: 2-hydroxy-6-oxo-4-(trifluoromethyl)-1H-pyridine-3-carbonitrile
Molecular Weight (g/mol): 204.11
Molecular Formula: C7H3F3N2O2
InChi Key: JRLZWXISQMMITA-UHFFFAOYSA-N
SMILES: OC1=C(C#N)C(=CC(=O)N1)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results