accessibility menu, dialog, popup

UserName

CAS RN 33208-99-0

NH2(S)ONH2H3CHCl

CAS RN 33208-99-0

IUPAC Name: 2-aminopropanamide hydrochloride
Synonyms: 2-aminopropanamide hydrochloride
Molecular Weight (g/mol): 124.57
Molecular Formula: C3H9ClN2O
InChi Key: FIAINKIUSZGVGX-UHFFFAOYNA-N
SMILES: Cl.CC(N)C(N)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)