CAS RN 330-14-3
CAS RN 330-14-3
IUPAC Name:
4-[(trifluoromethyl)sulfanyl]benzoyl chloride
Synonyms:
4-[(trifluoromethyl)sulfanyl]benzoyl chloride
Molecular Weight (g/mol):
240.62
Molecular Formula:
C8H4ClF3OS
InChi Key:
BCMFTOCCLOAGBP-UHFFFAOYSA-N
SMILES:
FC(F)(F)SC1=CC=C(C=C1)C(Cl)=O
1
–
3
of
3
results
1
4-(Trifluoromethylthio)benzoyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(Trifluoromethylthio)benzoyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(Trifluoromethylthio)benzoyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.