accessibility menu, dialog, popup

UserName

CAS RN 325850-81-5

OOHSNOO

CAS RN 325850-81-5

IUPAC Name: 2-[(3-{2,4-dioxo-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaen-3-yl}propyl)sulfanyl]benzoic acid
Synonyms: 2-[(3-{2,4-dioxo-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaen-3-yl}propyl)sulfanyl]benzoic acid
Molecular Weight (g/mol): 391.44
Molecular Formula: C22H17NO4S
InChi Key: GMVZUCHUOYUMLL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1SCCCN1C(=O)C2=CC=CC3=CC=CC(C1=O)=C23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results