accessibility menu, dialog, popup

UserName

CAS RN 3250-74-6

NNNNN

CAS RN 3250-74-6

IUPAC Name: 5-(pyridin-3-yl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(pyridin-3-yl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 146.13
Molecular Formula: C6H4N5
InChi Key: OFLOUIASTOSYPU-UHFFFAOYSA-N
SMILES: [N-]1N=NC(=N1)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results