CAS RN 3222-56-8
CAS RN 3222-56-8
IUPAC Name:
2-methylpyridine-3-carboxylic acid
Synonyms:
2-methylpyridine-3-carboxylic acid
Molecular Weight (g/mol):
137.14
Molecular Formula:
C7H7NO2
InChi Key:
HNTZKNJGAFJMHQ-UHFFFAOYSA-N
SMILES:
CC1=C(C=CC=N1)C(O)=O
1
–
1
of
1
results
1
2-Methylnicotinic acid, 98+%
CAS: 3222-56-8 Molecular Formula: C7H7NO2 Molecular Weight (g/mol): 137.14 MDL Number: MFCD00013449 InChI Key: HNTZKNJGAFJMHQ-UHFFFAOYSA-N Synonym: 2-methylnicotinic acid,2-methyl-nicotinic acid,2-methylnicotinicacid,3-pyridinecarboxylic acid, 2-methyl,2-methyl nicotinic acid,2-methyl-3-pyridinecarboxylic acid,methylnicotinic acid,pubchem2458,2-methylnicotinoic acid,acmc-1ctxp PubChem CID: 643373 IUPAC Name: 2-methylpyridine-3-carboxylic acid SMILES: CC1=C(C=CC=N1)C(=O)O