accessibility menu, dialog, popup

UserName

NH2HNFFF

CAS RN 3107-34-4

IUPAC Name: [2-(trifluoromethyl)phenyl]hydrazine
Synonyms: [2-(trifluoromethyl)phenyl]hydrazine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: JSWQDLBFVSTSIW-UHFFFAOYSA-N
SMILES: NNC1=CC=CC=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results