CAS RN 3080226-63-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris BioScience™ VVD 844
Potent covalent inhibitor of the RAS domain of the PI3K/p110α interaction