CAS RN 3067-19-4
IUPAC Name:
2-acetamido-3-methylbutanoic acid
Synonyms:
L-valine, N-acetyl-
Molecular Weight (g/mol):
159.19
Molecular Formula:
C7H13NO3
InChi Key:
IHYJTAOFMMMOPX-UHFFFAOYNA-N
SMILES:
CC(C)C(NC(C)=O)C(O)=O
1
–
3
of
3
results
1
N-Acetyl-DL-Valine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Acetyl-DL-Valine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Acetyl-DL-Valine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.