CAS RN 3053-17-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.