accessibility menu, dialog, popup

UserName

NaNaOSOONHNOHNNOSOOO(E)

CAS RN 3051-11-4

IUPAC Name: disodium 5-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-2-[(1E)-2-{4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-2-sulfonatophenyl}ethenyl]benzene-1-sulfonate
Synonyms: disodium 5-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-2-[(1E)-2-{4-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-2-sulfonatophenyl}ethenyl]benzenesulfonate
Molecular Weight (g/mol): 624.55
Molecular Formula: C26H18N4Na2O8S2
InChi Key: RDJCIKZLXHKBPH-SEPHDYHBSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC(NN=C2C=CC(=O)C=C2)=CC=C1\C=C\C1=CC=C(NN=C2C=CC(=O)C=C2)C=C1S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)