accessibility menu, dialog, popup

UserName

OHIINOO

CAS RN 305-85-1

IUPAC Name: 2,6-diiodo-4-nitrophenol
Synonyms: phenol, 2,6-diiodo-4-nitro-
Molecular Weight (g/mol): 390.9
Molecular Formula: C6H3I2NO3
InChi Key: UVGTXNPVQOQFQW-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(C=C1I)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results