accessibility menu, dialog, popup

UserName

CAS RN 3029-30-9

NN

CAS RN 3029-30-9

IUPAC Name: naphthalene-1,4-dicarbonitrile
Synonyms: naphthalene-1,4-dicarbonitrile
Molecular Weight (g/mol): 178.19
Molecular Formula: C12H6N2
InChi Key: BENSWQOUPJQWMU-UHFFFAOYSA-N
SMILES: N#CC1=C2C=CC=CC2=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)