CAS RN 300766-53-4
IUPAC Name:
4-methylbenzen-1-ide; iodozincylium
Synonyms:
4-methylbenzen-1-ide; iodozincylium
Molecular Weight (g/mol):
283.42
Molecular Formula:
C7H7IZn
InChi Key:
AHAJMZHMDYRVRB-UHFFFAOYSA-M
SMILES:
[Zn+]I.CC1=CC=[C-]C=C1