CAS RN 300364-84-5
IUPAC Name:
2-[3-(4-tert-butylphenyl)-2-methylprop-2-en-1-ylidene]propanedinitrile
Synonyms:
2-[3-(4-tert-butylphenyl)-2-methylprop-2-en-1-ylidene]propanedinitrile
Molecular Weight (g/mol):
250.35
Molecular Formula:
C17H18N2
InChi Key:
OIASAVWSBWJWBR-UHFFFAOYSA-N
SMILES:
CC(C=C(C#N)C#N)=CC1=CC=C(C=C1)C(C)(C)C
1
–
3
of
3
results
1
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.