accessibility menu, dialog, popup

UserName

CAS RN 2993-24-0

NH2SFFFFF

CAS RN 2993-24-0

IUPAC Name: 4-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 4-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 219.17
Molecular Formula: C6H6F5NS
InChi Key: MZGZUHNSMNNSRJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results