CAS RN 29843-90-1
CAS RN 29843-90-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Neodymium(III) bromide hydrate, REacton™, 99.9% (REO)
CAS: 29843-90-1 Molecular Formula: Br3Nd Molecular Weight (g/mol): 383.95 MDL Number: MFCD00748137 InChI Key: LBWLQVSRPJHLEY-UHFFFAOYSA-K IUPAC Name: neodymium(3+) tribromide SMILES: [Br-].[Br-].[Br-].[Nd+3]