CAS RN 298-04-4
IUPAC Name:
O,O-diethyl {[2-(ethylsulfanyl)ethyl]sulfanyl}phosphonothioate
Synonyms:
frumin caswell no. 341 glebofos disyston rcra waste number P039 ethyl thiometon ethylthiometon B di-syston G demeton (substance) insyst-D
Molecular Weight (g/mol):
274.39
Molecular Formula:
C8H19O2PS3
InChi Key:
DOFZAZXDOSGAJZ-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)SCCSCC
1
–
6
of
6
results