accessibility menu, dialog, popup

UserName

H3C(S)OHOHO(E)(Z)

CAS RN 29623-28-7

IUPAC Name: (9Z,11E,13S)-13-hydroxyoctadeca-9,11-dienoic acid
Synonyms: 13-hydroxyoctadecadienoic acid 13-hode 13-hodd
Molecular Weight (g/mol): 296.45
Molecular Formula: C18H32O3
InChi Key: HNICUWMFWZBIFP-IRQZEAMPSA-N
SMILES: CCCCC[C@H](O)\C=C\C=C/CCCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results