CAS RN 296-35-5
IUPAC Name:
1,4,7,10,13,16-hexaazacyclooctadecane
Synonyms:
1,4,7,10,13,16-hexaazacyclooctadecane
Molecular Weight (g/mol):
258.41
Molecular Formula:
C12H30N6
InChi Key:
RVJABZUDCPZPPM-UHFFFAOYSA-N
SMILES:
C1CNCCNCCNCCNCCNCCN1
1
–
3
of
3
results
1
Hexacyclen, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Hexacyclen, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Hexacyclen, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.