accessibility menu, dialog, popup

UserName

CH3BrOCH3

CAS RN 29578-83-4

IUPAC Name: 1-bromo-3-methoxy-5-methylbenzene
Synonyms: 1-bromo-3-methoxy-5-methylbenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: AOEVRCZZWJWKPG-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results