CAS RN 2925590-71-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ tri-GalNAc-PEG3-Azide
A Tocris product, part of R&D Systems. Asialoglycoprotein receptor ligands with PEG3 linker and azide group for onward chemistry