accessibility menu, dialog, popup

UserName

CAS RN 29211-70-9

CH3NNHO

CAS RN 29211-70-9

IUPAC Name: 3-propyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 5-propyl-2,4-dihydropyrazol-3-one
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: GVUNLYBSNQOHBD-UHFFFAOYSA-N
SMILES: CCCC1=NNC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results