CAS RN 2915-90-4
IUPAC Name:
didodecyl(methyl)amine
Synonyms:
didodecyl(methyl)amine
Molecular Weight (g/mol):
367.71
Molecular Formula:
C25H53N
InChi Key:
UWHRNIXHZAWBMF-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCN(C)CCCCCCCCCCCC
1
–
3
of
3
results
1
Didodecylmethylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Didodecylmethylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Didodecylmethylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.