CAS RN 2913-97-5
IUPAC Name:
2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetaldehyde
Synonyms:
phthalimide, N-(formylmethyl)-
Molecular Weight (g/mol):
189.17
Molecular Formula:
C10H7NO3
InChi Key:
LMRDBJZQDUVCQH-UHFFFAOYSA-N
SMILES:
O=CCN1C(=O)C2=CC=CC=C2C1=O
1
–
2
of
2
results
1
Phthalimidoacetaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Phthalimidoacetaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.