CAS RN 291275-45-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-Chloro-3-[(1Z)-1-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1-propen-1-yl]-benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Chloro-3-[(1Z)-1-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1-propen-1-yl]-benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Chloro-3-[(1Z)-1-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1-propen-1-yl]-benzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.