accessibility menu, dialog, popup

UserName

NH4OSOOHN

CAS RN 28836-03-5

IUPAC Name: ammonium 8-(phenylamino)naphthalene-1-sulfonate
Synonyms: ammonium 8-(phenylamino)naphthalene-1-sulfonate
Molecular Weight (g/mol): 316.38
Molecular Formula: C16H16N2O3S
InChi Key: IPBNQYLKHUNLQE-UHFFFAOYSA-N
SMILES: [NH4+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)