accessibility menu, dialog, popup

UserName

H3COOOCl

CAS RN 2881-63-2

IUPAC Name: ethyl 3-(4-chlorophenyl)-3-oxopropanoate
Synonyms: ethyl 3-(4-chlorophenyl)-3-oxopropanoate
Molecular Weight (g/mol): 226.66
Molecular Formula: C11H11ClO3
InChi Key: DGCZHKABHPDNCC-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)