accessibility menu, dialog, popup

UserName

CH3SNBr

CAS RN 28771-82-6

IUPAC Name: 5-bromo-4-methyl-2-phenyl-1,3-thiazole
Synonyms: 5-bromo-4-methyl-2-phenyl-1,3-thiazole
Molecular Weight (g/mol): 254.15
Molecular Formula: C10H8BrNS
InChi Key: XNYKJKJMPCGEQI-UHFFFAOYSA-N
SMILES: CC1=C(Br)SC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results