accessibility menu, dialog, popup

UserName

CAS RN 287172-74-1

HOOFFCl

CAS RN 287172-74-1

IUPAC Name: 2-chloro-3,6-difluorobenzoic acid
Synonyms: 2-chloro-3,6-difluorobenzoic acid
Molecular Weight (g/mol): 192.55
Molecular Formula: C7H3ClF2O2
InChi Key: COWJYTLVSFKOPM-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results