accessibility menu, dialog, popup

UserName

CH3NNCH3H2NH3C

CAS RN 28466-21-9

IUPAC Name: trimethyl-1H-pyrazol-4-amine
Synonyms: trimethylpyrazol-4-amine
Molecular Weight (g/mol): 125.18
Molecular Formula: C6H11N3
InChi Key: SSDGMKHZMNTWLS-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(N)=C1C
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)