accessibility menu, dialog, popup

UserName

CH3SOOHOHOHOH

CAS RN 28244-98-6

IUPAC Name: 2-(hydroxymethyl)-6-[(4-methylphenyl)sulfanyl]oxane-3,4,5-triol
Synonyms: 2-(hydroxymethyl)-6-[(4-methylphenyl)sulfanyl]oxane-3,4,5-triol
Molecular Weight (g/mol): 286.34
Molecular Formula: C13H18O5S
InChi Key: IQCLIQLFPVKINX-UHFFFAOYNA-N
SMILES: CC1=CC=C(SC2OC(CO)C(O)C(O)C2O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results