accessibility menu, dialog, popup

UserName

CAS RN 2820-37-3

CH3NHNH3C

CAS RN 2820-37-3

IUPAC Name: 4,5-dimethyl-1H-pyrazole
Synonyms: 3,4-dimethyl-2H-pyrazole
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: VQTVFIMEENGCJA-UHFFFAOYSA-N
SMILES: CC1=C(C)C=NN1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)