CAS RN 2809-69-0
CAS RN 2809-69-0
IUPAC Name:
hexa-2,4-diyne
Synonyms:
2,4-hexadiyne
Molecular Weight (g/mol):
78.11
Molecular Formula:
C6H6
InChi Key:
PCTCNWZFDASPLA-UHFFFAOYSA-N
SMILES:
CC#CC#CC
1
–
1
of
1
results
1
2,4-Hexadiyne, 98%, Thermo Scientific™
CAS: 2809-69-0 Molecular Formula: C6H6 Molecular Weight (g/mol): 78.11 MDL Number: MFCD00041617 InChI Key: PCTCNWZFDASPLA-UHFFFAOYSA-N Synonym: 2,4-hexadiyne,dimethylbutadiyne,dimethyldiacetylene,acmc-209h0v,ch3c.$.cc.$.cch3,hexa-2,4-diyn-1-yl PubChem CID: 137727 IUPAC Name: hexa-2,4-diyne SMILES: CC#CC#CC