accessibility menu, dialog, popup

UserName

CAS RN 280559-30-0

BOOH3CCH3H3CH3CCH3

CAS RN 280559-30-0

IUPAC Name: 4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 246.16
Molecular Formula: C15H23BO2
InChi Key: GTZNZJQZUWIDTN-UHFFFAOYNA-N
SMILES: CC(CB1OC(C)(C)C(C)(C)O1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results