accessibility menu, dialog, popup

UserName

CAS RN 27131-40-4

NHOClBr

CAS RN 27131-40-4

IUPAC Name: N-(2-bromophenyl)-4-chlorobutanamide
Synonyms: N-(2-bromophenyl)-4-chlorobutanamide
Molecular Weight (g/mol): 276.56
Molecular Formula: C10H11BrClNO
InChi Key: WIKLTDDDTXZFHP-UHFFFAOYSA-N
SMILES: ClCCCC(=O)NC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results