accessibility menu, dialog, popup

UserName

CAS RN 265108-38-1

H3CONH

CAS RN 265108-38-1

IUPAC Name: 4-(ethoxymethyl)piperidine
Synonyms: 4-(ethoxymethyl)piperidine
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: GXKNDCQREJHGRQ-UHFFFAOYSA-N
SMILES: CCOCC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results