accessibility menu, dialog, popup

UserName

IHNN

CAS RN 261953-36-0

IUPAC Name: 6-iodo-1H-indazole
Synonyms: 6-iodo-1H-indazole
Molecular Weight (g/mol): 244.04
Molecular Formula: C7H5IN2
InChi Key: RSGAXJZKQDNFEP-UHFFFAOYSA-N
SMILES: IC1=CC=C2C=NNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results