CAS RN 261951-78-4
CAS RN 261951-78-4
IUPAC Name:
1-fluoro-2-methoxy-4-(trifluoromethyl)benzene
Synonyms:
1-fluoro-2-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
194.13
Molecular Formula:
C8H6F4O
InChi Key:
AXEAQZIYCQTHQV-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC=C1F)C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-5-(trifluoromethyl)anisole, 97%
CAS: 261951-78-4 Molecular Formula: C8H6F4O Molecular Weight (g/mol): 194.129 MDL Number: MFCD01631564 InChI Key: AXEAQZIYCQTHQV-UHFFFAOYSA-N Synonym: 2-fluoro-5-trifluoromethyl anisole,1-fluoro-2-methoxy-4-trifluoromethyl benzene,4-fluoro-3-methoxybenzotrifluoride,pubchem8275,acmc-1ceet,4-fluoro-3-methoxy-benzotrifluoride,1-fluoranyl-2-methoxy-4-trifluoromethyl benzene PubChem CID: 2774771 IUPAC Name: 1-fluoro-2-methoxy-4-(trifluoromethyl)benzene SMILES: COC1=C(C=CC(=C1)C(F)(F)F)F