accessibility menu, dialog, popup

UserName

CAS RN 261901-57-9

CH3NNSOOHOHCl

CAS RN 261901-57-9

IUPAC Name: 3-({2-[2-(4-methylpiperidin-1-yl)ethyl]pyrrolidin-1-yl}sulfonyl)phenol hydrochloride
Synonyms: 3-{2-[2-(4-methylpiperidin-1-yl)ethyl]pyrrolidin-1-ylsulfonyl}phenol hydrochloride
Molecular Weight (g/mol): 388.95
Molecular Formula: C18H29ClN2O3S
InChi Key: XQCJOYZLWFNDIO-UHFFFAOYNA-N
SMILES: Cl.CC1CCN(CCC2CCCN2S(=O)(=O)C2=CC=CC(O)=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results