accessibility menu, dialog, popup

UserName

CAS RN 25569-97-5

IUPAC Name: thiophene-2-carbonyl thiophene-2-carboxylate
Synonyms: thiophene-2-carbonyl thiophene-2-carboxylate
Molecular Weight (g/mol): 238.28
Molecular Formula: C10H6O3S2
InChi Key: BKBCDDYQJQGMNX-UHFFFAOYSA-N
SMILES: O=C(OC(=O)C1=CC=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results