accessibility menu, dialog, popup

UserName

CAS RN 24439-00-7

NNNCl

CAS RN 24439-00-7

IUPAC Name: 6-chloro-2H,3H-imidazo[1,2-b]pyridazine
Synonyms: 6-chloro-2H,3H-imidazo[1,2-b]pyridazine
Molecular Weight (g/mol): 155.59
Molecular Formula: C6H6ClN3
InChi Key: KSDBTFCPAJCBSO-UHFFFAOYSA-N
SMILES: ClC1=NN2CCN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results