CAS RN 2418-95-3
CAS RN 2418-95-3
IUPAC Name:
(2S)-2-azaniumyl-6-{[(tert-butoxy)carbonyl]amino}hexanoate
Synonyms:
(2S)-2-ammonio-6-[(tert-butoxycarbonyl)amino]hexanoate
Molecular Weight (g/mol):
246.31
Molecular Formula:
C11H22N2O4
InChi Key:
VVQIIIAZJXTLRE-QMMMGPOBSA-N
SMILES:
CC(C)(C)OC(=O)NCCCC[C@H]([NH3+])C([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals N(epsilon)-Boc-L-lysine, 97%
CAS: 2418-95-3 Molecular Formula: C11H22N2O4 Molecular Weight (g/mol): 246.31 MDL Number: MFCD00037221 InChI Key: VVQIIIAZJXTLRE-QMMMGPOBSA-N Synonym: h-lys boc-oh,lys boc,nepsilon-tert-butoxycarbonyl-l-lysine,n6-boc-l-lysine,n∼6∼-tert-butoxycarbonyl-l-lysine,nepsilon-boc-l-lysine,ne-boc-l-lysine,n epsilon-boc-l-lysine,h-lys boc-2-chlorotrityl resin,n6-tert-butoxycarbonyl-l-lysine PubChem CID: 2733283 IUPAC Name: (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid SMILES: CC(C)(C)OC(=O)NCCCC[C@H]([NH3+])C([O-])=O