accessibility menu, dialog, popup

UserName

CH3NH3COCl

CAS RN 24167-52-0

IUPAC Name: 3-chloro-N,N-dimethylbenzamide
Synonyms: 3-chloro-N,N-dimethylbenzamide
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: JSXDHOISXRKLGG-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC(Cl)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results