accessibility menu, dialog, popup

UserName

CAS RN 2393-17-1

NH2OHO

CAS RN 2393-17-1

IUPAC Name: 3-(4-aminophenyl)propanoic acid
Synonyms: 4-aminohydrocinnamic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: WXOHKMNWMKZMND-UHFFFAOYSA-N
SMILES: NC1=CC=C(CCC(O)=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)