accessibility menu, dialog, popup

UserName

NH2NSNHNHN

CAS RN 23771-52-0

IUPAC Name: 4-amino-1H,2H,6H-pyrazolo[3,4-d]pyrimidine-6-thione
Synonyms: 4-amino-1H,2H-pyrazolo[3,4-d]pyrimidine-6-thione
Molecular Weight (g/mol): 167.19
Molecular Formula: C5H5N5S
InChi Key: YJMNLDSYAAJOPX-UHFFFAOYSA-N
SMILES: NC1=NC(=S)N=C2NNC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results