accessibility menu, dialog, popup

UserName

CAS RN 23144-52-7

HOCl

CAS RN 23144-52-7

IUPAC Name: 8-chlorooctan-1-ol
Synonyms: 8-chlorooctan-1-ol
Molecular Weight (g/mol): 164.67
Molecular Formula: C8H17ClO
InChi Key: YDFAJMDFCCJZSI-UHFFFAOYSA-N
SMILES: OCCCCCCCCCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results