accessibility menu, dialog, popup

UserName

H3COBrSOOCl

CAS RN 23095-05-8

IUPAC Name: 5-bromo-2-methoxybenzene-1-sulfonyl chloride
Synonyms: 5-bromo-2-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 285.54
Molecular Formula: C7H6BrClO3S
InChi Key: IXSBNNRUQYYMRM-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Br)C=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)