CAS RN 23052-81-5
CAS RN 23052-81-5
IUPAC Name:
(2S)-2-azaniumyl-4-phosphonatobutanoate
Synonyms:
L-AP4
Molecular Weight (g/mol):
181.09
Molecular Formula:
C4H8NO5P
InChi Key:
DDOQBQRIEWHWBT-VKHMYHEASA-L
SMILES:
[NH3+][C@@H](CCP([O-])([O-])=O)C([O-])=O
1
–
2
of
2
results
1
(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals
CAS: 23052-81-5 Molecular Formula: C4H10NO5P Molecular Weight (g/mol): 183.1 MDL Number: MFCD00083244 InChI Key: DDOQBQRIEWHWBT-VKHMYHEASA-N Synonym: l-ap4,2s-2-amino-4-phosphonobutanoic acid,l-+-2-amino-4-phosphonobutyric acid,chembl33567,s-2-amino-4-phosphonobutyrate,l-ap-4,3h ap4,l-2-amino-4-phosphonobutiric acid,l-1-amino-4-phosphonobutanoic acid,s-2-amino-4-phosphonobutanoic acid PubChem CID: 179394 IUPAC Name: (2S)-2-amino-4-phosphonobutanoic acid SMILES: C(CP(=O)(O)O)C(C(=O)O)N
2
Tocris Bioscience™ L-AP4
CAS: 23052-81-5 Molecular Formula: C4H10NO5P